Anti-ID4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-6669R-A488
BOSSBS-6669R-A488EA 893 EUR
BOSSBS-6669R-A488
Anti-ID4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Members of the Id family of basic helix-loop-helix (bHLH) proteins include Id1 (1–3), Id2 (4), Id3 and Id4 (5). They are ubiquitously expressed and dimerize with members of the class A and B HLH proteins (1–5). Due to the absence of the basic region, the resulting heterodimers cannot bind DNA. The Id-type proteins thus appear to negatively regulate DNA binding of bHLH proteins. Since Id1 inhibits DNA binding of E12 and Myo D, it apparently functions to inhibit muscle-specific gene expression. Under conditions that facilitate muscle cell differentiation, the Id protein levels fall,More Product Information
Order Now

SPECIFICATIONS

Antigen symbol ID4
Environmentally Preferable
Antigen name Inhibitor Of Differentiation 4
Antigen synonyms bHLHb27|IDB4
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 488
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 3400
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR