Anti-CCDC5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-6908R-A750
BOSSBS-6908R-A750EA 893 EUR
BOSSBS-6908R-A750
Anti-CCDC5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
HAUS1 is 1 of 8 subunits of the 390-kD human augmin complex, or HAUS complex. The augmin complex was first identified in Drosophila, and its name comes from the Latin verb 'augmentare,' meaning 'to increase.' The augmin complex is a microtubule-binding complex involved in microtubule generation within the mitotic spindle and is vital to mitotic spindle assembly (Goshima et al., 2008 [PubMed 18443220]; Uehara et al., 2009 [PubMed 19369198]).[supplied by OMIM, Jun 2010].

Type: Primary
Antigen: CCDC5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/278
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR