Anti-GDF2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Supplier: Bioss
BS-6909R-A350
BOSSBS-6909R-A350EA 893 EUR
BOSSBS-6909R-A350
Anti-GDF2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in rodents suggest that this protein plays a role in the adult liver and in differentiation of cholinergic central nervous system neurons. [provided by RefSeq, Jul 2008].