Anti-DEFA6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-7129R-FITC
BOSSBS-7129R-FITCEA 1350 EUR
BOSSBS-7129R-FITC
Anti-DEFA6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Has very low antimicrobial activity against Gram-negative and Gram-positive bacteria. May protect cells against infection with HIV-1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DEFA6
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR