Anti-ADGRB1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-7513R-A350
BOSSBS-7513R-A350EA 893 EUR
BOSSBS-7513R-A350
Anti-ADGRB1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
BAI 1 protein is likely to be a potent inhibitor of angiogenesis in brain and may play a significant role as a mediator of the p53 signal in suppression of glioblastoma. It may function in cell adhesion and signal transduction in the brain. Reduced or no expression is observed in some glioblastoma cell lines and cancer tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADGRB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol ADGRB1
Environmentally Preferable
Antigen name Adhesion G Protein-Coupled Receptor B1
Antigen synonyms GDAIF|BAI1|R75078|KIAA4089|Brain-Specific Angiogenesis Inhibitor 1
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 350
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 575
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR