Anti-CLK3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-7908R-A350
BOSSBS-7908R-A350EA 893 EUR
BOSSBS-7908R-A350
Anti-CLK3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Phosphorylates serine- and arginine-rich (SR) proteins of the spliceosomal complex. May be a constituent of a network of regulatory mechanisms that enable SR proteins to control RNA splicing. Phosphorylates serines, threonines and tyrosines.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLK3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol CLK3
Antigen name CDC-like kinase 3
Environmentally Preferable
Antigen synonyms PHCLK3/152|PHCLK3
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 350
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 1198
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR