Anti-ABHD14B Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-7945R-A680
BOSSBS-7945R-A680EA 893 EUR
BOSSBS-7945R-A680
Anti-ABHD14B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
ABHD14B has hydrolase activity towards p-nitrophenyl butyrate (in vitro). It may activate transcription. There are 2 isoforms produced by alternative splicing.

Type: Primary
Antigen: ABHD14B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 122-210/210
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR