Anti-ASNA1 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-7972R
BOSSBS-7972REA 725 EUR
BOSSBS-7972R
Anti-ASNA1 Rabbit Polyclonal Antibody
Antibodies
ASNA1 is the human homolog of the E.coli arsA gene which is an ATPase, and is the catalytic component of a multisubunit oxyanion pump responsible for resistance to arsenicals and antimonials.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ASNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR