Anti-CDK2AP2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-7995R-CY7
BOSSBS-7995R-CY7EA 900 EUR
BOSSBS-7995R-CY7
Anti-CDK2AP2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
This gene encodes a protein that interacts with cyclin-dependent kinase 2 associated protein 1. Pseudogenes associated with this gene are located on chromosomes 7 and 9. Alternatively spliced transcript variants have been observed for this gene. [provided by RefSeq, Dec 2012].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDK2AP2
Clonality: Polyclonal
Clone: 0
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR