Anti-CCDC88B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Supplier: Bioss
BS-8141R-A647
BOSSBS-8141R-A647EA 893 EUR
BOSSBS-8141R-A647
Anti-CCDC88B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
HkRP3, also known as CCDC88B (coiled-coil domain-containing protein 88B) or BRLZ (brain leucine zipper domain-containing protein), is a 1,476 amino acid protein that belongs to the CCDC88 family. Members of the hook-related protein family are characterized by the presence of a C-terminal hook-related domain and an N-terminal potential microtubule binding domain. HkRP3 may be involved in the linkage of various organelles to microtubules, and exists as six alternatively spliced isoforms. The gene encoding HkRP3 maps to human chromosome 11q13.1 and mouse chromosome 19 A. Chromosome 11 housesMore Product Information