Anti-NPFF Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-8150R-A647
BOSSBS-8150R-A647EA 893 EUR
BOSSBS-8150R-A647
Anti-NPFF Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
FMRFamide related peptides are morphine modulating peptides that have many physiologic effects, including the modulation of morphine induced analgesia, elevation of arterial blood pressure, and increased somatostatin secretion from the pancreas. Neuropeptide FF potentiates and sensitizes ACCN2 and ACCN3 channels.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NPFF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR