Anti-NPFF Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-8150R-HRP
BOSSBS-8150R-HRPEA 900 EUR
BOSSBS-8150R-HRP
Anti-NPFF Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
FMRFamide related peptides are morphine modulating peptides that have many physiologic effects, including the modulation of morphine induced analgesia, elevation of arterial blood pressure, and increased somatostatin secretion from the pancreas. Neuropeptide FF potentiates and sensitizes ACCN2 and ACCN3 channels.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: NPFF
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR