Anti-FAHD2A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-8228R-A680
BOSSBS-8228R-A680EA 893 EUR
BOSSBS-8228R-A680
Anti-FAHD2A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
The FAH family contains two highly homologous 314 amino acid proteins, designated FAHD2A (fumarylacetoacetate hydrolase domain-containing protein 2A) and FAHD2B (fumarylacetoacetate hydrolase domain-containing protein 2A). FAHD2A and B utilise calcium and magnesium as cofactors, and may possess hydrolase activity. The genes encoding FAHD2A/B map to human chromosome 2, the second largest human chromosome, which consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2. HarlequinMore Product Information