Anti-GIMAP4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-8270R-A350
BOSSBS-8270R-A350EA 893 EUR
BOSSBS-8270R-A350
Anti-GIMAP4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Exhibits intrisinic GTPase activity. Shows a higher affinity for GDP over GTP (about 12-fold higher), and binding shows an absolute requirement for magnesium.Tissue specificity:Highly expressed in spleen and peripheral blood leukocytes that contain mostly T- and B-lymphocytes. Expressed specifically in resting T- and B-lymphocytes and expression significantly decreases during B- or T-lymphocyte activation. Expressed at lower levels in thymus, ovary, colon and small intestine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GIMAP4
Clonality: Polyclonal
Clone:
Conjugation:More Product Information
Order Now

SPECIFICATIONS

Antigen symbol GIMAP4
Antigen name GTPase IMAP Family member 4
Antigen synonyms IAN1|IMAP4|MSTP062|IAN-1
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 350
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 55303
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR