Anti-CPXM1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-8341R-CY5
BOSSBS-8341R-CY5EA 900 EUR
BOSSBS-8341R-CY5
Anti-CPXM1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
CPXM (carboxypeptidase X, member 1) belongs to the peptidase M14 family. However, no carboxypeptidase activity has yet been detected. It may be involved in cell-cell interactions.Members of the M14 metallocarboxypeptidase protein family serve many diverse functions and are divided into three subfamilies based on structure, function and amino acid sequence similarity. Belonging to the N/E subfamily, CPXM (metallocarboxypeptidase CPX-1) is a 734 amino acid protein that contains a F5/8 type C domain and likely binds one zinc ion per subunit. Most members of the N/E subfamily contain severalMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CPXM1
Environmentally Preferable
Antigen name Carboxypeptidase X (M14 Family), Member 1
Antigen synonyms CPX1|CPXM
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 56265
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR