Anti-FGFR1OP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-8345R-A750
BOSSBS-8345R-A750EA 893 EUR
BOSSBS-8345R-A750
Anti-FGFR1OP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
FOP, is a 399 amino acid protein that localises to the centrosome and contains one LisH domain. Expressed ubiquitously with highest expression in kidney, heart, muscle, colon, liver, testis and pancreas, FOP functions as a homodimer that interacts with EB1 and CEP350 and is essential for anchoring microtubules to centrosomes. Chromosomal aberrations that involve the FOP gene are associated with the pathogenesis of stem cell myeloproliferative disorder (MPD), a condition that is characterised by eosinophilia and myeloid hyperplasia and ultimately leads to acute myeloid leukemia. FOP isMore Product Information