Anti-IP3 Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-8869R-A750
BOSSBS-8869R-A750EA 893 EUR
BOSSBS-8869R-A750
Anti-IP3 Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Intracellular channel that mediates calcium release from the endoplasmic reticulum following stimulation by inositol 1,4,5-trisphosphate. Plays a role in ER stress-induced apoptosis. Cytoplasmic calcium released from the ER triggers apoptosis by the activation of CaM kinase II, eventually leading to the activation of downstream apoptosis pathways (By similarity).

Type: Primary
Antigen: IP3 Receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/2758
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR