Anti-SPTLC2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-9027R-A488
BOSSBS-9027R-A488EA 893 EUR
BOSSBS-9027R-A488
Anti-SPTLC2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
This gene encodes a long chain base subunit of serine palmitoyltransferase. Serine palmitoyltransferase, which consists of two different subunits, is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine. Mutations in this gene were identified in patients with hereditary sensory neuropathy type I. [provided by RefSeq, Mar 2011].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPTLC2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR