Anti-ACCSL Rabbit Polyclonal Antibody

Supplier: Bioss
BS-9062R
BOSSBS-9062REA 1090 EUR
BOSSBS-9062R
Anti-ACCSL Rabbit Polyclonal Antibody
Antibodies
Belongs to the class-I pyridoxal-phosphate-dependent aminotransferase family.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ACCSL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR