Anti-MURF3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-9376R-A750
BOSSBS-9376R-A750EA 893 EUR
BOSSBS-9376R-A750
Anti-MURF3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
MuRF1, is a nuclear protein that interacts with SMT3b and the large myofibrillar protein Titin. In muscle cells, MuRF2 (RFN29) regulates gene expression and protein turnover. It localises to the cytoplasm, but under atrophic conditions it is detected in the nucleus. MuRF2 can form oligomers with various other proteins, including Titin and Myosin. MuRF3, also designated tripartite motif-containing 54 (TRIM54) or ring finger protein 30 (RNF30), interacts with tubulin and stabilises microtubules duing myotube formation. It is a cytoplasmic protein the localises to the Z-lines in skeletalMore Product Information
Order Now

SPECIFICATIONS

Antigen name MURF3
Environmentally Preferable
Antigen synonyms Muscle-specic RING finger protein 3|Tripartite mot containing 54|MURF 3|tibody Tripartite mot-containing protein 54|Muscle specic RING finger protein homolog|TRIM54|RNF30|MuRF3|Ring finger protein 30|MURF-3|MURF|RNF28|Muscle specic RING finger protein 3|TRIM 54|Muscle specic RING finger protein|Muscle-specic RING finger protein|TRI54_HUMAN
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 750
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 57159
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Epitope 131-240/358
Immunogen KLH conjugated synthetic peptide derived from human MURF3
Form Liquid
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR