Anti-CNOT2 Ser101 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-9384R-A680
BOSSBS-9384R-A680EA 1350 EUR
BOSSBS-9384R-A680
Anti-CNOT2 Ser101 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
CNOT2 (CCR4-NOT transcription complex subunit 2) is a ubiquitous protein encoded by the human gene CNOT2. CNOT2 belongs to the CNOT2/3/5 family and is part of the CCR4-NOT complex. The CCR4-NOT complex is an evolutionarily conserved, multi-component complex known to be involved in transcription as well as mRNA degradation. Various subunits (e.g. CNOT1, CNOT2) are involved in influencing nuclear hormone receptor activities. The CCR4-NOT complex is also involved in the regulation of Histone H3 lysine 4 methylation through a ubiquitin-dependent pathway that likely involves the proteasome.More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name CNOT2 Ser101
Antigen synonyms NOT2 negative regulator of transcription 2 yeast homolog|HSPC131|CCR4-associated factor 2|p-CNOT2Ser101|Negative regulator of transcription 2|CCR4 associated factor 2|CCR4 NOT transcription complex subunit 2|CDC36|CNOT2 phospho S101|CNOT2|MSTP046|CNOT2_HUMAN|NOT2H|CCR4-NOT transcription complex subunit 2|NOT2
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 4848
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Phosphorylation (Ser101)
Immunogen KLH conjugated synthetic phosphopeptide derived from human CNOT2 around the phosphorylation site of Ser101
Form Liquid
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR