Anti-CARKD Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Supplier: Bioss
BS-9402R-A647
BOSSBS-9402R-A647EA 893 EUR
BOSSBS-9402R-A647
Anti-CARKD Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
CARKD is a 347 amino acid protein that belongs to the YjeF family and exists as three alternatively spliced isoforms. Containing one YjeF C-terminal domain, CARKD is encoded by a gene that maps to human chromosome 13q34. Chromosome 13 contains around 114 million base pairs and 400 genes. Key tumor suppressor genes on chromosome 13 include the breast cancer susceptibility gene, BRCA2, and the RB1 (retinoblastoma) gene. RB1 encodes a crucial tumor suppressor protein which, when defective, leads to malignant growth in the retina and has been implicated in a variety of other cancers. The geneMore Product Information