Anti-SLC25A37 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-9523R-A647
BOSSBS-9523R-A647EA 893 EUR
BOSSBS-9523R-A647
Anti-SLC25A37 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Mitoferrin1, SLC25A37, belongs to the mitochondrial carrier family and contains 3 Solcar repeats. It is a mitochondrial iron transporter that specifically mediates iron uptake in developing erythroid cells and plays an essential role in heme biosynthesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC25A37
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR