Anti-Mitoferrin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-9523R-A680
BOSSBS-9523R-A680EA 893 EUR
BOSSBS-9523R-A680
Anti-Mitoferrin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Mitoferrin1, SLC25A37, belongs to the mitochondrial carrier family and contains 3 Solcar repeats. It is a mitochondrial iron transporter that specifically mediates iron uptake in developing erythroid cells and plays an essential role in heme biosynthesis.

Type: Primary
Antigen: Mitoferrin 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/338
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR