Anti-ASH1L Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-9671R-CY5
BOSSBS-9671R-CY5EA 893 EUR
BOSSBS-9671R-CY5
Anti-ASH1L Rabbit Polyclonal Antibody (Cy5®)
Antibodies
ASH1L is a 2,969 amino acid protein encoded by the human gene ASH1L. ASH1L belongs to the histone-lysine methyltransferase family (SET2 subfamily) and contains three AT hook DNA-binding domains, one AWS domain, one BAH domain, one bromodomain, one PHD-type zinc finger, one post-SET domain and one SET domain. It is a widely expressed nuclear protein with highest expression found in brain, heart and kidney. ASH1L is a histone methyltransferase and is believed to methylate 'Lys-4' of Histone H3, which is a specific tag for epigenetic transcriptional activation.
Recommended Dilutions: IF(IHC-More Product Information