Anti-C20orf11 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-9689R-A488
BOSSBS-9689R-A488EA 893 EUR
BOSSBS-9689R-A488
Anti-C20orf11 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
TWA1 is a 228 amino acid nuclear protein that is highly conserved throughout evolution. TWA1 interacts with Ran BP-M, a protein that is implicated in a diverse array of cellular processes including DNA replication, entry into and exit from mitosis, and the transport of RNA and proteins through the nuclear pore complex. Together, Ran BP-M and TWA1 form a complex with cytoplasmic Muskelin, a mediator of cell spreading. It is thought that this complex plays a role in the Ran GTPase cycle and therefore a potential role in the cell cycle. TWA1 contains a LisH-CTLH motif which is usually found inMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol C20orf11
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 488
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR