Anti-NOL1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-9710R-CY5.5
BOSSBS-9710R-CY5.5EA 900 EUR
BOSSBS-9710R-CY5.5
Anti-NOL1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Proliferation-associated nucleolar antigen is expressed in mid G1 phase with peak level during the S phase and a rapid degradation during late mitosis. Its expression in breast carcinoma is correlated with patient prognosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOL1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR