Anti-B4GALT7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-9728R-A647
BOSSBS-9728R-A647EA 893 EUR
BOSSBS-9728R-A647
Anti-B4GALT7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
β-1,4-galactosyltransferases (β-1,4-Gal-T) are type II membrane-bound glycoproteins that are substrate-specific and function to transfer galactose in a β-1,4 linkage to an acceptor sugar. There are seven members of the β-1,4-Gal-T family, all of which are directed to the golgi apparatus through a hydrophobic sequence at the N-terminus. β-1,4-Gal-T7, also known as B4GALT7 or XGALT1, is a 327 amino acid single-pass type II membrane protein that is expressed at high levels in heart, pancreas and liver. β-1,4-Gal-T7 uses manganese to catalyze the UDP-dependent biosynthesis ofMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol B4GALT7
Antigen name xylosylprotein beta 1,4-galactosyltransferase, polypeptide 7
Environmentally Preferable
Antigen synonyms EDSP1|XGALT1|XGPT1
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 647
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 11285
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR