Brady® R6400 Printer Ribbon for BBP11 and BBP12 Label Printers
Supplier: Brady
112648
BRDY112648EA 111 EUR
BRDY112648
Brady® R6400 Printer Ribbon for BBP11 and BBP12 Label Printers
Printer Ribbons
This ink-side-out ribbon features an open 12,7 mm core and is to be used for the Brady BBP11 label printer and BBP12 label printer.
  • One-colour ribbon for printing a single colour onto a label
  • R6400 series resin formulation offers superior chemical and abrasion resistance when printed on the appropriate label material from Brady®
  • Combine brady label materials with their recommended ribbon series for superior legend performance
Recommended for use with materials designed to resist chemicals, such as B-492 frozen surface polyester and B-481 chemicalMore Product Information