Labels for BBP33 printers, type B-437
Supplier: Brady
133846 133847 133888 133889 133980 133981 133990 133997 133791
BRDY133846EA 514 EUR
BRDY133846 BRDY133847 BRDY133888 BRDY133889 BRDY133980 BRDY133981 BRDY133990 BRDY133997 BRDY133791
Labels for BBP33 printers, type B-437
Labels
Durable B-437 topcoated polyvinylfluoride material is used most frequently in wire and cable identification applications where self-extinguishing features are required.
  • White or yellow labels feature a matte finish and are thermal transfer printable
  • Service temperature, –70 to 135 °C
This material utilises a permanent acrylic adhesive and has excellent resistance to water, oil and other common fluids.
Certifications: Meets the requirements of MIL-M-87958, pressure sensitive adhesive wire or cable marker and Identification specification; UL approved; CSA accepted.