Brady® Polyester Label Tape for BradyJet™ J7300 Colour Label Printer
Supplier: Brady
170452 176577 176624 176625 176600 176578
BRDY170452EA 643 EUR
BRDY170452 BRDY176577 BRDY176624 BRDY176625 BRDY176600 BRDY176578
Brady® Polyester Label Tape for BradyJet™ J7300 Colour Label Printer
Labels
General-purpose polyester labels from Brady feature a specially formulated topcoat for improved durability, colour, and appearance.
  • Inkjet-receptive polyester material showcases vibrant colours, sharp text, and precise graphics
  • Enhanced topcoat works with durable J7300 inks to protect printed text and graphics from smudging and harsh conditions
  • Aggressive adhesive
  • Designed for indoor/outdoor use
Inkjet-receptive polyester labels adhere strongly to smooth surfaces for a wide range of applications, and are durable up to two years outdoors, even longerMore Product Information