PermaSleeve® High Temp Heat-Shrink Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Supplier: Brady
173556 173563 173558 173557 173546 173573 173569 173560 173575 173562 173572 173551 173570 173566 173549 173568 173547 173559 173574 173543 173552 173545 173567 173565 173576 173550 173571 173554 173564 173548 173577
BRDY173556EA 287 EUR
BRDY173556 BRDY173563 BRDY173558 BRDY173557 BRDY173546 BRDY173573 BRDY173569 BRDY173560 BRDY173575 BRDY173562 BRDY173572 BRDY173551 BRDY173570 BRDY173566 BRDY173549 BRDY173568 BRDY173547 BRDY173559 BRDY173574 BRDY173543 BRDY173552 BRDY173545 BRDY173567 BRDY173565 BRDY173576 BRDY173550 BRDY173571 BRDY173554 BRDY173564 BRDY173548 BRDY173577
PermaSleeve® High Temp Heat-Shrink Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Labels
The B-345 PermaSleeve® material is suitable for high temperature and low outgassing applications, or applications that require greater resistance to harsh chemicals.
  • 2:1 heat shrinkable sleeves are fade-resistant and flame-retardant, and meet the requirements of NASA vacuum outgassing specification SP-R-0022A
  • sleeves can be used in applications that require greater resistance to harsh chemicals such as fuels, lubricants, and high power cleaning solvents
  • operating temperature range of –55 to +225 °C (–67 to +437 °F)
  • matte finish
Brady authenticMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable