Ink Cartridge for BradyJet™ J7300 Colour Label Printer, CMY
Supplier: Brady
176504
BRDY176504EA 170 EUR
BRDY176504
Ink Cartridge for BradyJet™ J7300 Colour Label Printer, CMY
Inks
Get better results with BradyJet™ J7300 inks. Designed specifically for the J7300, BradyJet™ ink cartridges produce vibrant colours, last long, and deliver value in every drop.
  • CMY ink cartridge delivers vibrant, full-colour labels that demand attention
  • Works in tandem with separate K True Black ink cartridge to mix more accurate, durable colours
  • For use with the BradyJet™ J7300 colour label printer
Ink works with enhanced J7300 label material topcoat to dry instantly and won’t smudge, smear, or spot. Self-contained cartridge drops into the printerMore Product Information