Label Printer, QWERTY UK with Brady Workstation PWID Suite, M610
Supplier: Brady
317806
BRDY317806EA 0 EUR
BRDY317806
Label Printer, QWERTY UK with Brady Workstation PWID Suite, M610
Printers Label Printers
Design and print labels using a keypad or desktop software.
  • Work longer with an enhanced Gen3 Li-ION battery - print 4,500 labels on a full charge
  • Easily import and export label files, lists and graphics to the printer through USB 2.0 port (Type A and B) or USB flash drive
  • Print labels at 50,80 mm per second print speed
  • Get added durability with a printer that is resistant to military-grade shocks (MIL-STD-810G) and 1,80 m drops
  • Operate your printer in –18 to +46 °C temperatures
Get the durability your work demands. Do more with enhanced capabilitiesMore Product Information
Order Now
SPECIFICATIONS
Compatibility Stand-alone and Peripheral
Environmentally Preferable
Materials supported Continuous and Die-cut
Model M610-QY-UK-PWID
Printer Monocolour
Print method Thermal transfer
Print resolution 300 dpi
Print speed 750 (max. labels per day)
Print width 50,80 mm (max.)