Label Printer, UK, M611
Supplier: Brady
317836
BRDY317836EA 0 EUR
BRDY317836
Label Printer, UK, M611
Printers Label Printers
Create and share label files to print from anywhere using Bluetooth or Wi-Fi.
  • Create and print up to 50,80 mm width labels
  • Print labels at 50,80 mm per second print speed
  • Design, preview and print any way you choose: from your phone, tablet, or computer
  • Store more than 1,000 labels on the printer, then print at the jobsite
  • Features a compact and lightweight design
  • Allows connectivity for up to 5 users via Bluetooth
Get more flexibility in the field, Design, Preview, Print. Any way you choose. Store more than 1,000 labels on board, access files easierMore Product Information