Labels, hazardous substance, GHS pictograms
Supplier: Brady
811678 811679 811680 811681 811682 811683 811684 811685 811686 811687 811688 811689 811690 811691 811692 811693 811694 811695 811696 811697 811698 811699 811700 811701 811702 811703 811704 811705 811706 811707 811708 811709 811710 811711 811712 811713 811714 811715 811716 811717 811718 811719 811720 811721 811722 814013 814014 814015 814016 814017 814018 814019 814020 814021 814022 814034 814033 814036 814037 814038 814039 814032 814035 814030 834181 834180 834182 834183 834187 834186 834184 834185 834188 814023 814024 814025 814026 814027 814029 814031 814028
116-0072EA 20.7 EUR
116-0072 116-0073 116-0074 116-0075 116-0076 116-0077 116-0078 116-0079 116-0080 116-0081 116-0082 116-0083 116-0084 116-0085 116-0086 116-0087 116-0088 116-0089 116-0090 116-0091 116-0092 116-0093 116-0094 116-0095 116-0096 116-0097 116-0098 116-0099 116-0250 116-0251 116-0252 116-0253 116-0254 116-0255 116-0256 116-0257 116-0258 116-0259 116-0260 116-0261 116-0262 116-0263 116-0264 116-0265 116-0266 116-0313 116-0314 116-0315 116-0316 116-0317 116-0318 116-0319 116-0320 116-0321 116-0322 BRDY814034 BRDY814033 BRDY814036 BRDY814037 BRDY814038 BRDY814039 BRDY814032 BRDY814035 BRDY814030 BRDY834181 BRDY834180 BRDY834182 BRDY834183 BRDY834187 BRDY834186 BRDY834184 BRDY834185 BRDY834188 BRDY814023 BRDY814024 BRDY814025 BRDY814026 BRDY814027 BRDY814029 BRDY814031 BRDY814028
Labels, hazardous substance, GHS pictograms
Labels
In line with the GHS (Global Harmonised System), identification elements need to be attached to chemical containers - in particular new warning symbols, hazard statements and safety data sheets. The new identification elements have been mandatory since 21 January 2009 and supplement the REACH regulation on the registration, evaluation, authorisation and restriction of chemicals.
  • Available in different sizes
  • Black and red on white
Order Now
SPECIFICATIONS
Environmentally Preferable