Graduated pipettes, SILBERBRAND ETERNA, type 2, Class B, total delivery
Supplier: Brand
27011 27013 27006 27009 27002 27005 27010
BRND27011EA 81.3 EUR
BRND27011 BRND27013 BRND27006 BRND27009 BRND27002 BRND27005 BRND27010
Graduated pipettes, SILBERBRAND ETERNA, type 2, Class B, total delivery
Pipettes Graduated Pipettes
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Nominal volume at the top
DIN EN ISO 835
Length: 360 mm ±10 mm except for 25 ml pipettes: 450 mm ±10 mm.
Certifications: DIN EN ISO 835
Caution: * Suitable for cotton plugs