Burette stopcocks, lateral or straight
Supplier: Brand
82120 82121 82122 82170 82220 82221 82222
BRND82120EA 59.8 EUR
BRND82120 BRND82121 BRND82122 BRND82170 BRND82220 BRND82221 BRND82222
Burette stopcocks, lateral or straight
Stopcocks
Borosilicate glass 3.3.
  • Interchangeable valve spindle
  • Spindle with retention device
  • Lateral or straight needle valve available
Valve housing with precision burette tip, PTFE spindle with PP cap.