Bulb pipettes (one mark), BLAUBRAND®, ETERNA, DAkkS
Supplier: Brand
DAKKS30507 DAKKS30512 DAKKS30514 DAKKS30515 DAKKS30518
BRNDDAKKS30507EA 92.2 EUR
BRNDDAKKS30507 BRNDDAKKS30512 BRNDDAKKS30514 BRNDDAKKS30515 BRNDDAKKS30518
Bulb pipettes (one mark), BLAUBRAND®, ETERNA, DAkkS
Pipettes Bulb Pipettes
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Supplied with DAkkS certificate
Certifications: DIN EN ISO 648; DE-M marking.
Order Now
SPECIFICATIONS
Environmentally Preferable