Volumetric flasks, class A, blue markings, DAkkS-calibrated, BLAUBRAND®
Supplier: Brand
DAKKS37259 DAKKS37258 DAKKS37291 DAKKS37289 DAKKS37290 DAKKS37243 DAKKS37246 DAKKS37238 DAKKS37255 DAKKS37248 DAKKS37249 DAKKS37250 DAKKS37251 DAKKS37252 DAKKS37253 DAKKS37254 DAKKS37247 DAKKS37263 DAKKS37236 DAKKS37245
BRNDDAKKS37259EA 114 EUR
BRNDDAKKS37259 BRNDDAKKS37258 BRNDDAKKS37291 BRNDDAKKS37289 BRNDDAKKS37290 BRNDDAKKS37243 BRNDDAKKS37246 BRNDDAKKS37238 BRNDDAKKS37255 BRNDDAKKS37248 BRNDDAKKS37249 BRNDDAKKS37250 BRNDDAKKS37251 BRNDDAKKS37252 BRNDDAKKS37253 BRNDDAKKS37254 BRNDDAKKS37247 BRNDDAKKS37263 BRNDDAKKS37236 BRNDDAKKS37245
Volumetric flasks, class A, blue markings, DAkkS-calibrated, BLAUBRAND®
Flasks Volumetric Flasks
Borosilicate glass 3.3.
  • Calibrated to contain (TC, In), incl. DAkkS-certificate
  • Mark and inscriptions in high contrast blue enamel
  • DE-M marking
*Wide neck.
Certifications: DIN EN ISO 1042
Order Now
SPECIFICATIONS
Environmentally Preferable