Anti-Endothelin B receptor Rabbit Polyclonal Antibody

Supplier: Biosensis
R-1550-100
BSENR-1550-100EA 321 EUR
BSENR-1550-100
Anti-Endothelin B receptor Rabbit Polyclonal Antibody
Antibodies
The Endothelin B receptor is a multi-pass membrane protein that belongs to the G-protein coupled receptor 1 family. It acts as a non-specific receptor for endothelin 1, 2, and 3.

Antibody detects ETRB at 51 kDa in WB in mouse fetus protein lysate. A weaker band at approx 75 kDa may also be observed (unknown).
Application Information:
WB. Suggested dilution of 1:1,000-1:5,000. Human Endothelin B receptor has a predicted length of 442 residues and MW of 50 kDa. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Endothelin BMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR