Amyloid plaque staining reagent, Amylo-Glo® RTD™
Supplier: Biosensis
TR-300-AG
BSENTR-300-AGEA 438 EUR
BSENTR-300-AG
Amyloid plaque staining reagent, Amylo-Glo® RTD™
Assays Cellular Assays
Amylo-Glo RTD reagent is designed to stain amyloid plaques in tissue sections.
  • For Amyloid plaques - Both intraneuronal and vascular
  • Very bright blue UV excitable stain under physiological conditions
  • Unique excitation/emission profile and mild staining
  • Combination for multiple immunofluorescent labeling studies
  • Storage Up to 6 months after date of receipt at 4 °C protected from light
Amylo-Glo RTD is compatible with fresh, frozen, and formalin-fixed immunohistochemistry or cytochemistry, and particularly good for confocal and multiple labeling because of its high fluorescent intensity and resistance to photo-bleaching. Moreover because Amylo-Glo® fluoresces in the UV channel, double and triple labeling experiments can be performed very easily.
Amylo-Glo® RTD™ is compatible with fresh, frozen, and formalin-fixed immunohistochemistry or cytochemistry, and it is particularly good for confocal and multiple labeling because of its high fluorescent intensity and high resistance to photo-bleaching. Moreover because Amylo-Glo® fluoresces in the UV channel, double and triple labeling experiments can be performed very easily - See protocol.