Anti-IgG Goat Polyclonal Antibody (CF™ 350)

Supplier: Biotium
20146 20146-1
20146.EA 238 EUR
20146. BTIU20146-1
Anti-IgG Goat Polyclonal Antibody (CF™ 350)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-rabbit IgG (H+L) with CF™350 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Blue fluorescent CF™350 (Ex/Em 347/448 nm) is more water-soluble, more photostable and at least 50% more fluorescent on proteins compared to the traditional blue dye AMCA.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF350
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR