Anti-ACP5 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ACP5/1070]

Supplier: Biotium
BNUM1070-50
BTIUBNUM1070-50EA 417 EUR
BTIUBNUM1070-50
Anti-ACP5 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ACP5/1070]
Antibodies
This antibody recognizes a protein of 35 kDa, which is identified as tartrate-resistant acid phosphatase (TRAcP). It exists as two isoforms (5a and 5b). This MAb reacts with both the isoforms. Serum TRAcP 5a is secreted by macrophages and dendritic cells and increased in many patients of rheumatoid arthritis.Serum TRAcP 5b is produced from osteoclasts and elevated during bone resorption. TRAcP is an iron containing glycoprotein, which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is the most basic of the acid phosphatases and is the only form notMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ACP5
Antigen name TRAcP
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone Clone ACP5/1070
Reactivity Human,Rat,Mouse
Host Mouse
Gene ID 1211
Isotype IgG2b kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Purification BSA-free
Concentration 1 mg/mL
Storage buffer PBS/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR