I-300 compatibility kit V-7X0
Supplier: BUCHI
11I300S40
BUCH11I300S40EA 2910 EUR
BUCH11I300S40
I-300 compatibility kit V-7X0
Evaporators Rotary Evaporators
Interface I-300 for use with BUCHI Rotavapour® R-21x/V-7x0 models.
  • Control all process parameters from single controller
  • Central control of all process parameters such as rotation speed, temperature (heating, cooling, vapour) and pressure
  • Remote monitoring on smart phones including push notifications and live view
The Interface I-300 controls all distillation parameters of a Rotavapour system. Modes: manual, timer, cont., AutoDest, drying. Mobile app is available.
Features are: Interface: I-300 Compatibility Pack: R-210/R-215, V-700/V-710.