Storage Boxes, Multipurpose
Supplier: BURKLE
3418-0129 3418-0148
216-1720EA 57 EUR
216-1720 BURK3418-0148
Storage Boxes, Multipurpose
Boxes Storage Boxes
Made of HDPE, these practical storage containers are ideal for safe transport of pourable or liquid contents.
  • With closed gripping edges
  • Can be stacked
  • Rounded inner edges and smooth inner surfaces for easy cleaning
  • Enables use in foodstuffs/hygiene sector