Stand for Imhoff funnels
Supplier: BURKLE
7118-2000
BURK7118-2000EA 136 EUR
BURK7118-2000
Stand for Imhoff funnels
Retort Stands
PP, chrome-plated steel. The stand for sedimentation funnels is used to keep, hold or store Imhoff funnels safely and reliably during sedimentation.
  • Adjustable height up to 450 mm
  • For funnels with Ø 90 mm or more
Two funnel holders ensure that Imhoff funnels will be held safely and securely and can be placed in an absolutely vertical position. Thanks to an innovative magnet concept no clamping screws are necessary. The height of the funnel holder can be adjusted easily and flexibly with one hand. With the corresponding accessory the holders for ImhoffMore Product Information