Tubing, silicone
Supplier: BURKLE
8803-0812 8803-0508 8803-0710 8803-0609
BURK8803-0812EA 54.3 EUR
BURK8803-0812 BURK8803-0508 BURK8803-0710 BURK8803-0609
Tubing, silicone
Tubing Transfer Tubing
Silicone tubing is a versatile transparent hose.
  • Expands while using organic solvents
  • Autoclavable, ethylene oxide
  • Appropriate for food
  • Withstand temperature from −60 to +180 °C
  • Good resistance against weak acids
To the extend it is non-reactive to ionising solvents except chlorinated aliphatic and aromatic hydrocarbons.
Hardness (Shore A): 55 ±5
Caution: Should not use it for harsh solutions and strong acids.
Order Now
SPECIFICATIONS
Environmentally Preferable