Anti-NFkB-p105 Rabbit Polyclonal Antibody [clone: A901]

Supplier: Bioworld Technology
BS1251
BWRLBS1251EA 283 EUR
BWRLBS1251
Anti-NFkB-p105 Rabbit Polyclonal Antibody [clone: A901]
Antibodies
Synthetic peptide, corresponding to amino acids 920-970 of Human NFkB-p105.

NFkB-p105 (A901) pAb detects endogenous levels of NF?B-p105 protein.This antibody does not recognize NFkB-p50 subunit.

Type: Primary
Antigen: NFkB-p105
Clonality: Polyclonal
Clone: A901
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR