Anti-PCNA Rabbit Polyclonal Antibody [clone: I88]

Supplier: Bioworld Technology
BS1289
BWRLBS1289EA 283 EUR
BWRLBS1289
Anti-PCNA Rabbit Polyclonal Antibody [clone: I88]
Antibodies
Synthetic peptide, corresponding to amino acids 51-100 of Human PCNA.

PCNA (I88) pAb detects endogenous levels of PCNA protein.

Type: Primary
Antigen: PCNA
Clonality: Polyclonal
Clone: I88
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR